Would you like to log in again?
Automation Pipette Tips for Tecan® Workstations, Porex BioProducts
Supplier: Porex
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
172-96R 171-96R BLKF17596RS1000 BLK-175-96R BLK-175-96RS 175-96RS BLK-172-96RS BLK-173-96RS 173-96R 175-96R 173-96RS 171-11 LMF172-96RS-50 LMF171-96RS-10 172-96RS
46300-408CS 904.4 USD InStock
46300-408 46300-398 46300-420 46300-388 46300-410 46300-440 46300-386 46300-428 46300-418 46300-404 46300-426 46300-424 46300-414 46300-422 46300-400
Automation Pipette Tips for Tecan® Workstations, Porex BioProducts
Pipette Tips
Tips are compatible with, but not limited to, the Tecan® Genesis workstation.
Volumes range from 10 to 1000 µl. Available in two different colors, natural and black. Also available with or without filter. The flexible polypropylene design provides optimal fit for the workstation. Sterile tips are tray-packaged.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty