Would you like to log in again?
Cap Strips for Tubes, Molecular Bio Products
Supplier: Molecular BioProducts
847-S 847-11
46300-542CS 204.29 USD InStock
46300-542 46300-540
Cap Strips for Tubes, Molecular Bio Products
Closures Test/Sample Tube Closures
Seal tubes tightly for storage
  • Made of polyethylene
  • For 0.75 and 1.2 mL sterile tubes
  • Available pre-sterilized as the material will not withstand autoclaving
Can be used with rows of tubes or cut apart for individual samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty