Would you like to log in again?
Nalgene® Dropper Bottles, Low-Density Polyethylene, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2751-9125 2751-9025 2752-9125 2750-9025 2753-9050 2750-9050 2750-9125 2752-9025 2751-9050 2752-9050
46300-596CS 64.22 USD InStock
46300-596 46300-594 46300-592 46300-608 46300-606 46300-604 46300-602 46300-614 46300-600 46300-598
Nalgene® Dropper Bottles, Low-Density Polyethylene, Thermo Scientific
Bottles Dropping and Dispensing Bottles
Drop volume averages 40 µl per drop, depending on reagent used. Both natural and white dropper bottles have excellent chemical resistance and have been rated suitable for most biotech, diagnostic, and pharmaceutical applications. Natural, clear dispensing bottles permit easy content identification, while white dispensing bottles protect UV light-sensitive products. Closures are polypropylene and are available in white or assorted colors (five each of white, red, yellow, green, blue) for easy reagent identification.
Dropper bottles provide accurate, repeatable dispensing of various reagents and offer an alternative to pipetting.
Ordering information: Each pack includes tips, closures, and bottles ready for assembly.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty