Would you like to log in again?
Orion™ AQUAfast™ Tests and Liquid Reagents, Thermo Scientific
Supplier: Thermo Scientific
Orion™
CODH00 CODL00 CODHP0 AC2009 AC2009
14232-922PK 85.02 USD InStock
14232-922 46300-630 14232-920 14232-924 CA46300-630
Orion™ AQUAfast™ Tests and Liquid Reagents, Thermo Scientific
Environmental Test Kits
Orion™ AQUAfast™ Digestion Tubes, Reaction Tubes, and Liquid Reagents are designed for use with Orion™ AQUAfast™ colorimeters and Orion™ AquaMate Vis and UV-Vis spectrophotometers.
Digestion and reaction tubes contain the precise amount of reagent, are made of special optical glass, and fit the Orion™ AQUAfast™ COD165 thermoreactor. Digestion tubes measure COD, total nitrogen, and total phosphate. Reaction tubes measure ammonia, nitrate, and phosphate. Liquid reagents mix quickly and easily in solution and measure fluoride (SPADN), hydrazine, and pH.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty