Would you like to log in again?
Orion™ AQUAfast II Chemistries Test Kits, Thermo Scientific
Supplier: Thermo Scientific
Orion™
AC2061 AC2001 AC2027 AC2007 AC2071 AC2046 AC2048 AC2095-WA AC2016 AC2012 AC2002 AC2099 AC2070 AC2029 AC2001 AC2030 AC2082 AC2060 AC2078 AC2096 AC2072 AC2065 AC2017 AC2055
46300-628PK 56.41 USD InStock
46300-628 46300-640 46300-662 46300-674 46300-652 46300-660 46300-650 46300-672 46300-670 CA46300-624 46300-648 46300-626 46300-624 46300-668 46300-658 46300-636 46300-644 46300-656 46300-634 46300-642 46300-664 46300-632 46300-676 46300-654
Orion™ AQUAfast II Chemistries Test Kits, Thermo Scientific
Environmental Test Kits
Orion™ AQUAfast Tablet Reagents perform colorimetric measurements of common water parameters with convenient, rapidly dissolving tablets.
  • For use with ORION™ AQUAfast AQ4000, ORION™ AQUAfast AQ3070, ORION™ AQUAfast AQ3700, and discontinued ORION™ AQ2 series meters and other colorimeters
  • 50 to 100 tests per kit
Use Orion™ AQUAfast Tablet Reagents with Orion™ AQUAfast colorimeters and Orion™ AquaMate Vis and UV-Vis spectrophotometers.
Test kits are designed for use with Orion™ AQUAfast AQ4000, Orion™ AQUAfast AQ3070, Orion™ AQUAfast AQ3700, and discontinued Orion™ AQ2 series meters and othercolorimeters.
These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging. They are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide, and zinc.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty