Would you like to log in again?
VWR STOPCOCK 8MM CS50
Supplier: VWR International
VWR* Stopcock, 8Mm Nominal O.D^ VWR* Stopcock, 8Mm Nominal O.D, Ideal For Use With Liquids And Gases Under Low Pressure (Operating Pressure 7.0 Psi) Or Low Vacuum, Hdpe Body And Polypropylene Tap Provide Smooth Turning Action Without The Use Of Lubricants
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR STOPCOCK 8MM CS50 46600-035 Case of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty