Would you like to log in again?
TRIlites® 994 Tri-Polymer Gloves, AdvanTech, Mapa Professionnel
Supplier: Mapa Professionnel
TRIlites®
34994016 34994017 34994026 34994027 34994029 34994028 34994018 34994019
46600-166CS 372.69 USD InStock
46600-166 32916-140 32916-144 32916-142 46600-160 46600-162 46600-164 32916-146
TRIlites® 994 Tri-Polymer Gloves, AdvanTech, Mapa Professionnel
Gloves
Unique tri-polymer blend of neoprene, nitrile, and natural rubber combines comfort with added strength, tear resistance, and chemical splash protection
  • Powder-free for no cross-contamination
  • Rolled cuff
  • Unlined
  • Ambidextrous
  • Color: Purple
Textured fingertips have a lightly halogenated finish for a high level of tactile sensitivity and grip in wet or dry conditions. Exclusive finish allows easy donning and glove layering, and helps keep tape and labels from sticking to gloves.
Certifications: Exceed ASTM D3578 AQL. In compliance with 21 CFR 170-199. CE marked.
Ordering information: Gloves are flat-packed 100 per sealed, printed bag or dispenser box.
Caution: This product contains natural rubber latex proteins, which may cause allergic reactions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty