Would you like to log in again?
Nalgene® Septum Closures, Sterile, Bulk Pack, Thermo Scientific
Supplier: Thermo Scientific
342178-0240 342178-0384
46600-596CS 526.08 USD InStock
46600-596 46600-597
Nalgene® Septum Closures, Sterile, Bulk Pack, Thermo Scientific
Closures Bottle Closures
Nalgene High Density Polyethylene closures with PTFE septum combines elastomeric sealing of silicone with non-reactive PFTE reagent contact surface
The closures are meant for use with Nalgene Sterile PET and PETG media bottles. They are non-cytotoxic, and non-pyrogenic, and have a dual septum material that provides improved gas barrier.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty