Would you like to log in again?
VWR GOGGLES DIRECT VENT STERL
Supplier: VWR International
VWR* Sterile Disposable Goggles^ Fog-free Exxene* coated lenses of 100% polycarbonate. Cleanroom manufactured, meet ANSI Z87.1 standards. Residuals nearly undetectable. Bio-burden: 1e-6. Weight: 80g. Direct venting with adjustable, non-linting rubber headband. Able to withstand up to 100[degree]C.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR GOGGLES DIRECT VENT STERL 46600-610 Case of 50 Retrieving
Each Retrieving
Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty