Would you like to log in again?
PYREX® Volumetric Pipettes, Class A, Serialized, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PYREX®
7103C-6 7103C-10 7103C-9 7103C-4 7103C-25 7103C-5 7103C-20 7103C-1 7103C-2 7103C-15 7103C-3 7103C-7 7103C-8
46610-106CS 232.49 USD InStock
46610-106 46610-116 46610-108 46610-118 46610-124 46610-102 46610-112 46610-126 46610-104 46610-114 46610-120 46610-122 46610-110
PYREX® Volumetric Pipettes, Class A, Serialized, Corning
Pipettes Graduated Pipettes
These PYREX® volumetric pipettes are calibrated to Class A tolerances in accordance with ASTM E-542 and ASTM E-969.
  • Reusable precision pipettes
  • Color-coded markings are enameled onto the glass
PYREX® serialized/certified volumetric glassware is now tested and calibrated in an ISO/IEC 17025 certified laboratory.
Each pipette is individually serialized and supplied with a Certificate of Identification and capacity, traceable to NIST standards. These precision PYREX® volumetric pipettes are manufactured and calibrated 'To Contain' (TC: bottom line) and 'To Deliver' (TD: top line) to specifications for Class A volumetric ware.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty