Would you like to log in again?
Flip-Top Containers, Capitol Vial®
Supplier: Capitol Vial
02FREGORED 03CLROYALBLUE 03CLGOLD 02WHITE 02EVERGREEN 02PINK 02YELLOW 02LTBROWN 02WINE 2.5CL 02AMBER 02PURPLE 03CLLTORANGE 03EDM33 02LTORANGE 03CLTURQ 02SKYBLUE 03CL 03CLYELLOW 02CL 02SILVER 03CLSKYBLUE 03CLFREGORED 03CLLIME 02BLACK 02TURQ 02ROYALBLUE 08CL 02LTRED 02DKORANGE 02GOLD 03CLPURPLE 02LIME
33500-728CS 244.37 USD InStock
33500-728 46610-138 46610-578 46610-556 46610-558 46610-134 46610-552 46610-554 46610-576 46610-136 46610-570 46610-152 46610-594 46610-572 46610-550 46610-132 46610-172 33999-042 46610-568 46610-546 46610-548 46610-564 46610-146 46610-542 46610-566 46610-588 46610-544 46610-142 46610-582 46610-164 46610-560 46610-144 46610-580
Flip-Top Containers, Capitol Vial®
Sample Containers General Purpose Containers
Triple-seal containers are airtight, leakproof, nonabsorptive, and nonaerosoling.
Unique process physically closes vials in the mold during manufacture to ensure an airtight seal and zero particulates >0.5µm. Sterile.
Made of FDA-approved, food-grade 100% polypropylene. Hinged top permits opening and closing these flip-top vials with one hand. Ideal for storing and packaging a wide range of solids and liquids in applications such as medical device packaging, reagent packaging, solid sample storage, and water testing.
Certifications: Vials conform to FDA and SAMHSA specifications.
Ordering information: Container 46610-588 features 10mL increments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty