Would you like to log in again?
Serum Vials, Borosilicate Glass, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
62113U-3 62121U-6 62113U-10 62413D-2 62121D-20 62113D-312 62113D-2 62421D-5 62121D-10 62121D-5 62113U-5 62421D-10 62421D-30 62121U-10 62113U-2
66013-127CS 882.02 USD InStock
66013-127 66013-129 66013-152 66013-150 66013-156 46610-208 66013-122 66013-143 66013-131 66013-120 46610-204 46610-206 46610-202 46610-210 89095-988
Serum Vials, Borosilicate Glass, Kimble Chase, DWK Life Sciences
Vials
Ideally suited for handling, containing, and storing a variety of liquids, including reagents, vaccines, and chromatography samples.
  • Autoclavable
These crimp-top serum vials are lighter in weight than molded bottles. More closely held tolerances and uniformity of glass allow the vials to be lyophilized.
Ordering information: Vials available in clear or amber glass, without closure. Stoppers and seals sold separately.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty