Would you like to log in again?
Protector® Standard Storage Cabinets, Labconco®
Supplier: Labcono
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Protector®
9900700 9904100 9900200 9900800 9904200 9900600 9904000 9900500 9904400 9900300 9900000 9904300 9904500 9900100 9900400
46610-418EA 2010 USD InStock
46610-418 46610-538 46610-406 46610-414 46610-402 46610-416 46610-448 46610-404 46610-454 46610-478 46610-400 46610-450 46610-452 46610-462 46610-470
Protector® Standard Storage Cabinets, Labconco®
Furniture Storage Cabinets
Standard cabinets support Labconco® laboratory hoods and can be used for storage of small laboratory instruments.
  • Available in six widths and two heights. Single-door models can be ordered with left or right hinge opening
Cabinets are constructed of white epoxy-coated steel and include one 20.3 cm (8") filler panel to increase cabinet’s overall depth from 55.9cm (22") to 76.2 cm (30"), for installation flush to wall.
Ordering information: Items designated as REDISHIP can be shipped within 48 hours of receiving your order. Cabinets are supplied with manual-closing, non-locking doors; four leveling feet; and one 20.3 cm (8") filler panel. Shelves (sold separately) are available for all cabinet widths. A work surface (also sold separately) is required for these cabinets, when used to support hoods without integral bottoms.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty