Would you like to log in again?
Flip-Top Containers with Lock Seal, Capitol Vial®
Supplier: Capitol Vial
04LPLS 10LSCL 04HPLS 02LS
46610-586CS 150.68 USD InStock
46610-586 46610-596 46610-584 46610-590
Flip-Top Containers with Lock Seal, Capitol Vial®
Sample Containers General Purpose Containers
Triple-seal containers are sterile, airtight, and leakproof
Unique process physically closes containers in the mold during manufacture to ensure an airtight seal and zero particulates >0.5µ. Process also produces sterile containers that do not require a second means of sterilization for most applications. Color: clear.
Nonabsorptive, nonaerosoling containers are made of FDA-approved, food grade 100% polypropylene. Hinged top enables opening and closing with one hand. Ideal for storing and packaging a wide range of solids and liquids in applications such as medical device packaging, reagent packaging, solid sample storage, and water testing. Containers feature a lock seal tab that allows the user to lock the lid to the base of the container.
Certifications: Containers conform to FDA and SAMHSA specifications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty