Would you like to log in again?
Minigrip® Red Line™ Reclosable Zip Bags
Supplier: Minigrip
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Minigrip®
MGRL2W0304 MGRL2P1315 MGRL4WH0406 MGRL4P1010 MGRL2P0303 MGRL4P1212 MGRL2P0508 MGRL4PH0508 MGRL4PH0404 MGRL2P0812 MGRL2P0406 MGRL2P1212 MGRL4P0609 MGRL2W0609 MGRL2P1012 MGRL4P1013 MGRL2P0404 MGRL2W0406 MGRL2P1215 MGRL4P0808 MGRL2P1013 MGRL4PH0304 MGRL4WH0609 MGRL4PH0606 MGRL4WH0810 MGRL2P0507 MGRL6P1012 MGRL4P0912 MGRL2P0408 MGRL6P0912 MGRL2P0808 MGRL4P1318 MGRL2P0608 MGRL2P0203 MGRL2W0810 MGRL2P0604 MGRL2P0912 MGRL2P0606 MGRL4PH0408 MGRL2P0304 MGRL2P0610 MGRL2P0810 MGRL6P0810 MGRL2P0505 MGRL4P1424 MGRL4WH0912 MGRL4PH02503 MGRL4PH0305 MGRL4P1215 MGRL4WH0508 MGRL2P0510 MGRL2P1318 MGRL2W0305 MGRL2P0305 MGRL4PH0406 MGRL2W0912 MGRL4P0810 MGRL4PH1012 MGRL6P0608 MGRL2P0609 MGRL4PH0608 MGRL2P0202 MGRL2W0203 MGRL4WH0305 MGRL2W0508
82027-714CS 237.03 USD InStock
82027-714 82027-716 82027-639 82027-710 82027-678 82027-712 82027-674 82027-676 46620-080 82027-680 82027-682 46620-001 46620-002 46620-005 46620-006 46620-008 83007-552 82027-649 83007-554 82027-688 82027-644 82027-646 82027-684 82027-640 82027-686 82027-643 82027-692 46610-644 82027-694 46620-012 82027-690 56766-380 83009-254 82027-699 82027-655 82027-656 56766-388 82027-658 82027-651 56766-385 82027-696 82027-653 56766-386 82027-660 46620-028 14221-792 46620-025 46620-026 82027-708 82027-704 82027-706 82027-666 82027-700 82027-668 82027-702 82027-662 82027-664 46620-030 82027-670 46300-564 82027-672 46300-562 46620-078 46620-032 46620-076
Minigrip® Red Line™ Reclosable Zip Bags
Bags Storage Bags
Re-closable low-density polyethylene bags provide easy-to-use, convenient packaging for all types of applications.
  • These are available with hang holes for ease in displaying items
  • Bags now feature a new double zipper closure for added security on bags 10" high or larger
Bags protect contents from dust, moisture, and other contaminants, and feature side seals to prevent splitting. They create an effective barrier without using much material or adding a significant amount of weight to the product. Bags are available with a white writing stripe to identify contents quickly. Bags accept most ink pens, labels, and hot stamps.
Ordering information: For additional sizes, please contact your Avantor sales representative.
Packaging: Most sizes are packed in DispenserBag® convenience packaging.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty