Would you like to log in again?
IsoClean™ Video- and Tape-Head Cleaner, MicroCare
Supplier: MicroCare
IsoClean®
MCC-BAC
46620-504CS 267.12 USD InStock
46620-504
IsoClean™ Video- and Tape-Head Cleaner, MicroCare
Soldering Supplies Flux Removers
Cear liquid IPA based flex remover that comes in a 12 oz aerosol. The ESD-Safe cleaner ensures easy removal of fluxes, pastes, greases and organic residues for a variety of items including circuits and stencils.
  • Cleans water-based Flux, rosin and no clean fluxes
  • Slow drying for maximum cleaning time
  • ESD-Safe
  • MicroCare Trigger Grip cleaning system compatible
Very effective on “no-clean” fluxes and pastes. Static- and plastic-safe. Mil-spec approved. Safe on delicate machinery.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty