Would you like to log in again?
WIPE FLUX REMOVER PRESATURATED PROCLEAN
Supplier: MicroCare
Proclean* Flux Remover Presaturated Wipe^ Proclean* Flux Remover Presaturated Wipe, Blend Of Three Ultra-Pure Alcohols, Cleans R, Ra, Rma, Oa, In No-Cleanin Fluxes & Solder Paste, Static-Safe, Medium Drying With Moderate Evaporation, Mild Alcohol Odor, Excellent Toxicity Ratings, Biodegradable
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WIPE FLUX REMOVER PRESATURATED PROCLEAN 46620-525 MCC-PROW Case of 1200 Retrieving
Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty