Would you like to log in again?
FLUX REMOVER SLOW-DRY CITRUS MINI 1GAL
Supplier: MicroCare
Bioact* Flux Remover^ 3.8L. Ozone-safe cleaner works well on rosin fluxes, synthetic fluxes, water-soluble fluxes and even some "no-clean" solders. Removes ionics, greases, oils, and contaminants. Also a screen cleaner and removes some conformal coatings. No rinsing.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FLUX REMOVER SLOW-DRY CITRUS MINI 1GAL 46620-664 MCC-EC7MG Each (1US Gal) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty