Would you like to log in again?
Formaldehyde-Free Rats
Supplier: VWR International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
140.1182.1 140.1178.1 140.1183.1 WARD470000-728 140.0952.10 WARD470000-732 140.1180.1 140.1179.1 140.1181.1 140.1182.10
470323-100EA 259.15 USD InStock
470323-100 470323-098 470000-736 470001-708 470323-102 470000-728 470323-112 470000-732 470323-096 470001-698
Formaldehyde-Free Rats
Specimens Preserved Specimens Preserved Animal Materials
Formaldehyde-free rats. Available in vacuum packs.
  • Mammalian anatomy
  • High quality preservation
To better serve the needs of all teaching environments and preferences, WARD’S offers this line as an alternative to our existing line of high-quality, safe preserved specimens. Absolutely no formaldehyde whatsoever is used in the fixing and preserving process for these specimens. Instead, our new solution used for preparation binds with the tissue as non-toxic, cross-linked proteins, making the solution truly 100% formaldehyde-free. WARD’S offers the largest selection of specimens prepared with this alternative process, including the widest range of sizes and injection styles. In addition, the specimens can be used comfortably over extended periods of time.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty