Would you like to log in again?
Freeze-Dried Bullfrogs
Supplier: VWR
WARD470027-850 WARD470001-404
470001-404PK 249.89 USD InStock
470001-404 470027-850
Freeze-Dried Bullfrogs
Specimens Preserved Specimens Preserved Animal Materials
Freeze-dried bullfrogs. Available in vacuum packs.
  • Amphibian anatomy
  • Economical
Our freeze-dried specimens, which use no chemicals, can be rehydrated in 10% alcohol for dissection. Size: 5" and over.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty