Would you like to log in again?
Ward's® Preserved Dogfish Sharks, 18 to 22"
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Ward's®
WARD470000-472 WARD470001-486 WARD470102-952 WARD470001-488 WARD470000-470 WARD470000-474
470000-472PK 139.99 USD InStock
470000-472 470000-474 470000-470 470102-952 470001-488 470001-486
Ward's® Preserved Dogfish Sharks, 18 to 22"
Specimens Preserved Specimens Preserved Animal Materials
Standard formalin preserved dogfish sharks.
  • Cartilaginous fish anatomy
  • No swim bladder or operculum
  • Gill slits
  • Good for group dissection
Dogfish sharks, Squalus sp., are a popular teaching specimen for biology and anatomy courses, specifically used for the teaching of cartilaginous fish anatomy. They are also great animal models to study comparative and vertebrate anatomy.
Dogfish will either be species Squalus sp. or Mustelus canis.
The harvesting of dogfish sharks is regulated by National Marine Fisheries to protect the species. Quantities used by our industry are very small in relation to the overall harvest.
Packaging: Packed in pails or vacuum-sealled bags.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty