Would you like to log in again?
Formaldehyde-Free Grassfrogs
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
136.0900.10 136.0904.10 136.0901.50 136.0913.100 136.0903.50 136.0899.50 136.0906.10 WARD470000-530 WARD470000-528 WARD470001-686
470001-682PK 106.4 USD InStock
470001-682 470000-528 470001-680 470000-524 470001-686 470000-526 470000-530 470001-676 470000-532 470000-522
Formaldehyde-Free Grassfrogs
Specimens Preserved Specimens Preserved Animal Materials
Formaldehyde-free grassfrogs. Available in pails or vacuum packs.
  • Amphibian anatomy
  • General biology
To better serve the needs of all teaching environments and preferences, WARD’S offers this line as an alternative to our existing line of high-quality, safe preserved specimens.
Absolutely no formaldehyde whatsoever is used in the fixing and preserving process for these specimens.
Instead, our new solution used for preparation binds with the tissue as non-toxic, cross-linked proteins, making the solution truly 100% formaldehyde-free. WARD’S offers the largest selection of specimens prepared with this alternative process, including the widest range of sizes and injection styles.
In addition, the specimens can be used comfortably over extended periods of time.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty