Would you like to log in again?
Laboratory Jacks
Supplier: United Scientific Supplies
LBJK01 LBJSET LBJSET
470001-938EA 145.25 USD InStock
470001-938 470006-698 470199-870
Laboratory Jacks
Laboratory Jacks
Laboratory support jack is constructed of painted aluminum and stainless steel.
  • Scissor jack design enables height adjustment with a simple turn of the knob
  • Sturdy design can support loads up to 20 kg
  • Platform height can be vertically adjusted from closed position of 2.75" up to 10.5"
  • Pre-drilled holes for attaching a support rod
Available as a set with removable support rod included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty