Would you like to log in again?
Ward's® Fixed 90 ̊ Photo Rules
Supplier: OREGON RULE CO.
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
4CSFWH3X3PLPGWARDS BSER10105 CSMWH8X8PLWARD
470350-688EA 2.25 USD InStock
470350-688 470001-982 470001-984
Ward's® Fixed 90 ̊ Photo Rules
Educational Materials Forensics Educational Materials Forensics Supplies
Easy to Measure at Right Angles
  • Easy to Read Photo Rules
  • Both English and Metric Scales
Precisely measure objects in two dimensions with these one-sided, "L-shaped", durable plastic rules. The English rule is 4" per side and is graduated in ¹⁄₁₆" increments; the metric rule is 11 cm per side and is graduated in 1 mm increments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty