Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Adjustable Leg Lab Stool

618A
470002-022EA 165 USD InStock
470002-022
Adjustable Leg Lab Stool
Chairs
These stools are adjustable and supported.

  • Eighteen-gauge tubular steel legs are supported by a ring to prevent wobbling
  • extendable legs to adjust the seat height to a comfortable level
  • Gray frame with a Masonite seat

Seat dia.: 14"
Weight: 10.5 lbs.
Adjustable steel legs from 19 - 27” (1” increments)

Ordering information: Assembly is required.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty