Would you like to log in again?
EZ-Open Pail
Supplier: Avantor
WARD470002-088
470002-088EA 27.95 USD InStock
470002-088
EZ-Open Pail
Buckets
EZ-Open pail with the gasketed lid.
  • Time saver and economical
  • Made of chemical-resistant polyethylene
  • Each pail has a plastic handle and a plastic hand grip
  • Aligned with GLOBE protocols
With just a few twists of wrist, lid can easily unscrewed and open these pails without needing a pail opener. Once after placing specimens in the pail, the lid can be tightly resealed to protect the contents for long-term storage.
Pail is labeled with the Ward’s Science logo.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty