Would you like to log in again?
Metabolism Experiment Kit
Supplier: LAB-AIDS INC (KITS) SE
Avg. Ratings: 1.0 (1 Review)
REVIEWS (1)
Total Ratings: 1
Avg. Ratings: out of 5
Sort all by:
55
470002-468KT 158 USD InStock
470002-468
Metabolism Experiment Kit
Educational Materials Biology Educational Materials Ecology Learning Activities
With the Metabolism Experiment Kit, students draw correlations between organism size, oxygen consumption, and temperature.
  • Time Required: One 50 Minute Class Period
  • Excellent Introduction to Metabolic Process
  • Teacher’s Guide with SDS Provided
Students measure the rate of oxygen consumption of small organisms such as insects, snails, worms, and germinating seeds. Using Patented (U.S. Pat. No. 3,581,737) LAB-AIDS® Metabolism chambers, they observe the organisms and draw correlations between the size of an organism and the temperature of the environment and oxygen consumption. They also calculate an organism’s respiratory rate in cubic cm of oxygen per hour, measure total gas change in a closed environment, and determine an organism’s metabolic relationship to total gas in a closed environment.
The kit includes two unbreakable chambers large enough for small animals and plants. One chamber is used to measure the organism's oxygen consumption, and the other serves as a control.
Ordering information: This kit includes metabolism chambers, carbon dioxide absorbent, capillary tubes with a connecting tube (calibrated), forceps, latticed organism holders with plastic retainers, index solution (colored, vial), student data cards, and a teacher’s guide with SDS. Animal or plant specimens for testing are needed but not included.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (1)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty