Would you like to log in again?
Chemical Equations Lab Activity
Supplier: AQUAPHOENIX SCIENTIFIC INC. SE
13-110
470002-480KT 161.45 USD InStock
470002-480
Chemical Equations Lab Activity
Educational Materials Chemistry Educational Materials Mixtures and Solutions Learning Activities
Introduce Chemical Equations Involving Single and Double Replacement Reactions
  • Ideal for Grades 8-12
  • Combine Science with Common Core Knowledge in one Lesson
Introduce students to chemical equations as they perform and observe two different single replacement reactions and two different double replacement reactions. Color changes, precipitate formation, and gas production are observed. The kit includes all required chemicals, a vocabulary exercise, and reproducible worksheets and laboratory procedures. Worksheets give students practice with chemical symbols, naming compounds, valence, and chemical formulas for compounds and radicals (polyatomic ions). Written laboratory exercises give students practice in identifying properties and completing and balancing single replacement and double replacement reactions. Laboratory supplies required.
Ordering information: Kit includes Instructor Manual, SDS, Zinc Pieces, Wood Splints, Aluminum Chloride, Ammonium Hydroxide, Sodium Hydroxide Solution, Hydrochloric Acid Solution, and Copper Sulfate Solution.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty