Would you like to log in again?
Chemical Properties Lab Activity
Supplier: AQUAPHOENIX SCIENTIFIC INC. SE
13-104
470002-482EA 281.55 USD InStock
470002-482
Chemical Properties Lab Activity
Educational Materials Chemistry Educational Materials Mixtures and Solutions Learning Activities
Test Chemical Properties of Seven Different Substances
  • Ideal for Grades 9-12
  • Chemical Properties in Microscale
Students test the chemical properties of seven different solutions in every combination and identify chemical properties by observing the reactions. Evidence of chemical reactions observed include gas production, color changes, and formation of precipitates. After becoming familiar with the chemical properties of the solutions, students use the information to identify any of four unknown solutions. The kit includes all required chemicals, reaction plates, reproducible worksheets an student procedures.
Ordering information: Kit includes Instructor's Manual, Special Needs Reference Sheet, SDS, Sodium bicarbonate, Hydrochloric Acid, Sodium Carbonate, Copper Sulfate, Ammonium Hydroxide, Phenolphthalein Indicator Solution, Acetic Acid, Sodium Hydroxide, Unknown A, Unknown B, Unknown C, Unknown D, Depression plates, Empty Dropper Bottles, and Labels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty