Would you like to log in again?
Denoyer-Geppert® Comprehensive Skin Model
Supplier: DENOYER-GEPPERT SCI CO SE
0185-00
470002-654EA 742.3 USD InStock
470002-654
Denoyer-Geppert® Comprehensive Skin Model
Models Human Biology Models
Details multiple skin conditions.
  • All Structures Enlarged
  • Guide Included
Healthy skin from the scalp, axilla, and palm or sole, are situated on one side. Represented structures include the subcutaneous layer with fat, dermis with dermal papillae, epidermis, hair and follicles, eccrine and apocrine sweat glands, sebaceous glands, blood vessels, Meissner’s and Pacinian corpuscles, nerves, goose bump muscles, and the erector pili that make hair stand on end. The other side of the model features examples of first, second, and third-degree burns, illustrating the increasing severity of damage to skin.
The model is mounted on a base and includes a key identifying 46 structures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty