Would you like to log in again?
Denoyer-Geppert® Giant Molar Tooth
Supplier: DENOYER-GEPPERT SCI CO SE
0107-00
470002-660EA 740.9 USD InStock
470002-660
Denoyer-Geppert® Giant Molar Tooth
Models Human Biology Models
Show Progressive Stages Of Caries With Various Inserts
  • 5 X Life Size
  • Removable Sections Of Temporal Bone
The most complete single tooth model we offer, the sectioned, 15X-enlarged upper molar with three roots can be dissected into seven different parts. Four interchangeable inserts allow you to alter the model from a healthy tooth to three increasingly advanced stages of a cavity; simply remove the pulp and a section of the crown and dentine to insert each section.
The model is mounted on a rotatable stand and can be removed for closer examination.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty