Would you like to log in again?
Vernier® Low-g Accelerometer
Supplier: Vernier
LGA-BTA
470002-950EA 98.99 USD InStock
470002-950
Vernier® Low-g Accelerometer
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Study one-dimensional acceleration.
  • Best choice for most acceleration experiments
The Low-g Accelerometer is the best choice for most acceleration experiments, including Newton’s second law, simple harmonic motion, and the relationship between acceleration and force. It can also be used to study one-dimensional motion of a car (real and toy), an elevator, a pendulum bob, or an amusement park ride.
Using Multiple Accelerometers, you can simultaneously measure the signals from two or three accelerometers mounted at right angles to measure acceleration in more than one dimension.
To study acceleration in a plane, mount two accelerometers at a right angle. This is a great way to study accelerations in an automobile.
Mount three accelerometers at right angles, or better yet use our 3-Axis Accelerometer, to study acceleration in every direction. This is useful for studying the complex motion of an amusement park ride or while bungee jumping.
Power: 30 mA at 5 VDC
Frequency Response: 0–100 Hz
For educational use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty