Would you like to log in again?
Vernier® Magnetic Field Sensor
Supplier: VERNIER SOFTWARE & TECH SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MG-BTA
470002-960EA 68.99 USD InStock
470002-960
Vernier® Magnetic Field Sensor
Electrodes Other Electrodes
Use to study magnetic fields.
  • Two ranges
This sensor uses a Hall effect transducer, and measures a vector component of the magnetic field near the sensor tip. It has two ranges, allowing for measurement of relatively strong magnetic fields around permanent magnets and electromagnets, as well as measurement of weak fields such as the Earth’s magnetic field. The articulated sensor tip allows you to measure both transverse and longitudinal magnetic fields.
For educational use only.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty