Would you like to log in again?
Vernier® Rotary Motion Sensor
Supplier: VERNIER SOFTWARE & TECH SE
RMV-BTD
470002-970EA 188.99 USD InStock
470002-970
Vernier® Rotary Motion Sensor
Electrodes Other Electrodes
Bidirectional angle sensor.
  • Measure rotational position, velocity, and acceleration
  • Measure linear position, velocity, and acceleration
  • Optical encoder: Bidirectional, quadrature encoder
  • Maximum speed: 30 rev/s at 1° resolution 7.5 rev/s at 0.25° resolution
  • 3-step pulley: 10, 29 and 48 mm groove diameter
The rotary motion sensor lets you monitor angular motion precisely and easily. It is direction sensitive. You can use it to collect angular displacement, angular velocity, and angular acceleration data. Typical experiments include measuring moments of inertia, torque, pendula, and Atwood’s machine experiments. Even though we call it a Rotary Motion Sensor, it can also be used to measure linear position to a fraction of a millimeter.
For educational use only.
Delivery information: It includes rotary motion sensor, thumb screw, 3-step pulley and mounting screw, O-ring.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty