Would you like to log in again?
Equilateral Prisms
Supplier: United Scientific Supplies
PGE050
470003-426EA 14.9 USD InStock
470003-426
Equilateral Prisms
Educational Materials Physics Educational Materials Optics Learning Activities
Easily demonstrate the separation of light.
  • Demonstrate reflection and refraction
  • Flint glass
Excellent quality, polished flint glass prisms for experiments in diffraction, dispersion, and spectrum formation. Individually boxed to prevent chipping.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty