Would you like to log in again?
Grafco® Sprague Rappaport Stethoscope
Supplier: GRAHAM-FIELD INC
Grafco®
602
470003-428EA 47 USD InStock
470003-428
Grafco® Sprague Rappaport Stethoscope
Stethoscopes
This professional, high-quality Sprague Rappaport type stethoscope has the best sound quality of all available stethoscopes.
  • Five interchangeable chest pieces (two diaphragms, three bells)
  • Ideal for infant, pediatric, and adult use
  • Heavy-walled 22" tubing decreases extraneous sounds
  • Latex-free
The exclusive, advanced design boasts an improved valve mechanism that virtually eliminates acoustic leakage, making it an excellent instrument to demonstrate heartbeat.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty