Would you like to log in again?
Generator, DC, Hand Held
Supplier: United Scientific Supplies
GENH02
470003-564EA 35.35 USD InStock
470003-564
Generator, DC, Hand Held
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Demonstrate the transfer of mechanical energy into electrical energy with a simple motion of your hand.
  • Transparent body
  • Comes with two 15" leads
The Hand Held DC Generator provides the means, you provide the power. Simply turn the hand-crank to produce a terminal voltage of 6.3V and a maximum current out of 0.2A. The transparent body lets everyone see exactly what is going on inside to produce this.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty