Would you like to log in again?
Fuel Cell Car Science Kit
Supplier: HORIZON FUEL CELL TECHNOLOGIES
FCJJ-11
470003-572EA 166.85 USD InStock
470003-572
Fuel Cell Car Science Kit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Use Clean Energy to Power a Car that You Build!
  • Components are visible during operation
  • Assembled car can steer itself
  • Includes an assembly manual
Experience the excitement of new fuel cell technology with this customized construction kit. In building this car, you will discover the possibility of using 100% clean fuel to power cars of the future. Once assembled, the car can steer itself independently and will reverse itself when it hits a barrier. A fully functioning vehicle, the car in this kit demonstrates the power of renewable energy on a small, user-friendly scale. A manual includes step by step instructions for simple assembly. All major car components are included - a fuel cell, a car chassis, a small electric motor, two water and gas storage containers, cables, tubes, a syringe, and more. All you need to add is water and two AA batteries.
WARNING: CHOKING HAZARD - Small parts. Not for children under 3 yrs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty