Would you like to log in again?
Fountain Connection Demonstration
Supplier: YEANY EDUCATIONAL PRODUCTS SE
10100
470003-680EA 7.75 USD InStock
470003-680
Fountain Connection Demonstration
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
This device will make you a 'Hero' in the classroom! Using two plastic pop bottles and this connection, you can demonstrate the fountain discovered by hero of Alexandria. By filling one of the bottles and inverting them you create a vacuum and send a stream of water upwards like a fountain.
The results appear to defy gravity and the laws of nature...but you know better!
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty