Would you like to log in again?
LaMotte® Acid Rain Study Kit
Supplier: LaMotte
3604-01
470003-688KT 94.8 USD InStock
470003-688
LaMotte® Acid Rain Study Kit
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Collect and Test Local Rainfall
  • Collect local rainwater samples
  • Measure water acidity
  • Includes a study guide
Your class of environmentalists will be completely equipped to test the rain in your area with this study outfit. They'll use a rain gauge with ground stake holder, pH kit measuring pH 3.0 - 6.5, octet comparator, and reagents. The kit comes packed in a waterproof plastic case with a stimulating study guide and suggested activities. It includes enough materials for 50 tests.
WARNING! This set contains chemicals that may be harmful if misused. Read cautions on individual containers carefully. Not to be used by children except under adult supervision.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty