Would you like to log in again?
Rocket Bulk Pack
Supplier: ESTES INDUSTRIES INC SE
1751 1756 1754 1755
470003-730CS 566.45 USD InStock
470003-730 470016-022 470006-266 470006-856
Rocket Bulk Pack
Educational Materials Physics Educational Materials Applied Physics Learning Activities
The Rocket Bulk Packs spark students' creativity.
  • Material for 12 Rockets
  • Students Improve Design and Building Skills
  • Additional Small Parts
  • Altitude Tracker
  • Technical Reports on Stability and Tracking
Student test their design and building skills.
Each pack contains enough parts to build 12 rockets as well as extra small parts.
Ordering information: This kit includes technical reports on stability and tracking and an altitude tracker for performance comparison. The following engines are recommended for use with this kit: 1/2A6-2, A8-3 (First Flight), B4-4, B6-6, C6-5, and C6-7.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty