Would you like to log in again?
Introduction to Conductivity Experiment Kit (Refill Kit Only)
Supplier: LAB-AIDS INC (KITS) SE
310
470003-766EA 258.45 USD InStock
470003-766
Introduction to Conductivity Experiment Kit (Refill Kit Only)
Educational Materials Chemistry Educational Materials Electrochemistry Learning Activities
With this kit students study the principles of conductors and insulators; study and develop the principles of electrolytes and non-electrolytes using acids, bases, salts, alcohols, and hydrocarbons; test a variety of common materials for their conductivity characteristics; and learn to differentiate between strong, weak, and non-conductors of electricity.
Ordering information: Three 9V batteries required for conductivity indicators, not included. Includes 3 LED conductivity indicators; 15 Lab-Aids® Chemplates® with spatula; 3 drop controlled btls deionized water; 2 drop controlled btls each 6M hydrochloric acid, denatured alcohol, glacial acetic acid; 1 vial each calcium hydroxide, sodium chloride crystals, sucrose crystals; 1 pkg. each copper wire with insulation, metal paper clips, plastic toothpicks, rubber bands, cut string; 30 student guides; 1 instruction manual with MSDS.
Caution: Note: Only the refill kit is available at this time.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty