Would you like to log in again?
Electric Plume
Supplier: SCIENCE INTERACTIVE GROUP
N-125
470003-972EA 42.9 USD InStock
470003-972
Electric Plume
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Graphically illustrate the fact that like charges repel each other.
The multi-colored ribbons are connected to a suction cup that sits on top of a Van de Graaff generator. As the generator operates, the ribbons accumulate a charge and stand on end.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty