Would you like to log in again?
Marble Ramp
Supplier: Avantor
WARD470003-992
470003-992KT 129.99 USD InStock
470003-992
Marble Ramp
Educational Classroom Kits and Activities
Ideal for students conduct experiments by varying the angle of the marble track as well as the height from which the marble is released.
  • Includes instructions
  • Grades 7 - 9
  • Size: 96.5 x 12.7cm
By gathering data and plotting a graph, students discover that the same result can often be achieved by different combinations of angle and height.
Caution: CHOKING HAZARD - Item contains a marble. Not for children under 3 yrs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty