Would you like to log in again?
Acrylic Prism, Set of 6
Supplier: United Scientific Supplies
PSET06
470004-004KT 76.05 USD InStock
470004-004
Acrylic Prism, Set of 6
Educational Materials Physics Educational Materials Optics Learning Activities
A durable and affordable collection of prisms.
  • Thick 1/2 " optical pieces
  • Includes and storage case and teacher manual
Made from clear acrylic sheet, this set contains six prisms in a variety of shapes. All sides are polished and the prisms all come in a protective wooden box with foam insert. It includes one rectangular block (3" x 2" x 1/2"), one semi-circle (3 1/2" base x 1/2" thick), one equilateral prism (3" sides, 1/2" thick), one double convex lens (3 1/2" long, 1" at center), two double concave lenses (3 1/2" long, 3/8" at center), and an activity guide.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty