Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Gastop for Teck™ Gas Jets
Supplier: TSUNAMI TECHNOLOGY GROUP INC.
4603100 GASTOP
470146-713BG 233.6 USD InStock
470146-713 470004-027
Gastop for Teck™ Gas Jets
Seals Gas Stops
Improve safety and prevent vandalism with these innovative caps, which lock on to most gas jets and can only be removed with the supplied key.
  • Gastop slips over hose barb, providing a vandal-proof seal
  • Seal against leakage
  • Prevent items from being inserted into gas jet
  • Unlocks with key
Designed to prevent accidental leaks, the caps are certified by the Canadian Gas Association and the American Gas Association. Please note: these gastops are made for use with Delta Faucet's Teck (tm) gas jets.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty