Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Gastop for Teck™ Gas Jets
Supplier: TSUNAMI TECHNOLOGY GROUP INC.
4603125 4603100 GASTOP
470004-026EA 14.2 USD InStock
470004-026 470146-712 470004-028
Gastop for Teck™ Gas Jets
Seals Gas Stops
Improve safety and prevent vandalism with these innovative caps, which lock on to most gas jets and can only be removed with the supplied key.
Designed to prevent accidental leaks, the caps are certified by the Canadian Gas Association and the American Gas Association. Please note: these gastops are made for use with Delta Faucet's Teck (tm) gas jets.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty