Would you like to log in again?
Field Soil Test Kit
Supplier: LUSTER LEAF PRODUCTS INC SE
1665
470004-044CS 1956.3 USD InStock
470004-044
Field Soil Test Kit
Standards Environmental Analysis Standards
Illustrate the Chemical Variety in Soil
  • Provides accurate and informative results
  • Includes recommendations for fertilizer use
  • Includes a teacher's guide
Students will test for nitrogen, phosphorus, potassium, and pH using syringes and test tubes to obtain accurate results. The supplied reference cards give information about each test, and offer recommendations for applying fertilizer. The kit contains enough reagents and materials for 120 tests and a teacher’s guide and comes in a sturdy carrying case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty