Would you like to log in again?
Ward's® Inheritance Patterns In Nasonia Lab Activity
Supplier: VWR
470004-172KT 207.69 USD InStock
470004-172
Ward's® Inheritance Patterns In Nasonia Lab Activity
Specimens Live Specimens Animals - Living Material
Investigate a unique reproductive system.
Designed for students with a more advanced knowledge of genetics, this sophisticated activity asks students to begin by determining if a gene is dominant, recessive, or sex-linked. Then they will design an experiment to test their hypothesis, including setting up a control experiment.
Once students are ready to collect their data, they will discover some perplexing facts that do not fit their hypothesis, and a Chi-square analysis should reject the sex-linked hypothesis. Eventually, students will come to realize that they are not dealing with a familiar genetic reproductive system. Instead, they are investigating a haplo-diploid system in which the males are haploid and the females are diploid.
This system will always mimic a sex-linked system since the males have only half the number of chromosomes as the females.
Ordering information: Kit contents include: Coupon redeemable for 3 Nasonia cultures and Sarcophaga pupae, 15 microviewing chambers, 50 Test tubes, 1 bag of cotton balls, 1 Working With Nasonia DVD, and Teacher's guide and student copymaster.
This item contains a coupon for live or perishable materials. At checkout, please provide the date you wish the perishable components to arrive.
Delivery information: The kit will ship separately from the live component. Kits do not contain perishable components and generally ship via UPS ground. Perishable components will ship via overnight or second day delivery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty