Would you like to log in again?
LaMotte® Leaf Pack Stream Ecology Lab Activity
Supplier: LaMotte
5882
470004-232EA 315 USD InStock
470004-232
LaMotte® Leaf Pack Stream Ecology Lab Activity
Educational Materials Biology Educational Materials Ecology Learning Activities
Study Freshwater Invertebrates and Their Ecosystem
  • Collect samples from a local stream
  • Practice tree and invertebrate identification
  • Make an artificial leaf pack
  • Aligned to NGSS 3-LS4-3, MS-LS2-1, MS-LS2-2, MS-LS2-4, MS-LS2-5, HS-LS2-1, HS-LS2-2, HS-LS2-7
Examine the habitat of forest streams and observe the interrelationship between the trees, food sources, aquatic organisms, and water quality. By making their own artificial “leaf packs”, students re-create the stream ecosystem, collecting its components, including leaves and freshwater macroinvertebrates, which they can compare with the included illustrations of leaf and invertebrate types for identification. It comes with enough materials for six setups.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty